3-Butylthiophene ≥97%

Supplier: Matrix Scientific

086494-25G
102848-898EA 763.6 USD
102848-898
3-Butylthiophene ≥97%
3-Butylthiophene

Hazard: Irritant

Formula: C₈H₁₂S
MW: 140.2459 g/mol
MDL Number: MFCD00143179
CAS Number: 34722-01-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR