trans-2-(2-Methoxybenzoyl)cyclohexane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086456-5G
102848-798EA 2307.69 USD
102848-798
trans-2-(2-Methoxybenzoyl)cyclohexane-1-carboxylic acid ≥95%
trans-2-(2-Methoxybenzoyl)cyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₈O₄
Melting Pt: 157…159 °C
MDL Number: MFCD01311265
CAS Number: 733742-72-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR