cis-2-Carbomethoxycyclohexane-1-carboxylic acid ≥97%

Supplier: Matrix Scientific

086447-5G
102848-744EA 496.92 USD
102848-744
cis-2-Carbomethoxycyclohexane-1-carboxylic acid ≥97%
cis-2-Carbomethoxycyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₉H₁₄O₄
Melting Pt: 67…70 °C
MDL Number: MFCD01311242
CAS Number: 111955-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR