cis-1,3-Cyclohexanedicarboxylic anhydride ≥97%

Supplier: Matrix Scientific

086446-1G 086446-10G
102848-736EA 490.77 USD
102848-736 102848-740
cis-1,3-Cyclohexanedicarboxylic anhydride ≥97%
cis-1,3-Cyclohexanedicarboxylic anhydride

Hazard: Irritant

Formula: C₈H₁₀O₃
Melting Pt: 188…190 °C
MDL Number: MFCD00077243
CAS Number: 4355-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR