trans-2-(2-Methoxybenzoyl)cyclopentane-1-carboxylic acid ≥95%
Supplier: Matrix Scientific
086442-5G
086442-1G
102848-712EA
2240
USD
102848-712
102848-716
trans-2-(2-Methoxybenzoyl)cyclopentane-1-carboxylic acid ≥95%
trans-2-(2-Methoxybenzoyl)cyclopentane-1-carboxylic acid
Hazard: Irritant
Formula:
C₁₄H₁₆O₄ Melting Pt: 96…99 °C |
MDL Number:
MFCD01311206 CAS Number: 733740-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...