cis-2-(2-Methoxybenzoyl)cyclopentane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086437-2G
102848-684EA 2115.38 USD
102848-684
cis-2-(2-Methoxybenzoyl)cyclopentane-1-carboxylic acid ≥95%
cis-2-(2-Methoxybenzoyl)cyclopentane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₆O₄
MDL Number: MFCD01311189
CAS Number: 732253-54-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR