cis-3-Benzoylcyclopentane-1-carboxylic acid ≥95%
Supplier: Matrix Scientific
086432-1G
086432-5G
102848-656EA
2172.31
USD
102848-656
102848-652
cis-3-Benzoylcyclopentane-1-carboxylic acid ≥95%
cis-3-Benzoylcyclopentane-1-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₄O₃ Melting Pt: 64…67 °C |
MDL Number:
MFCD01311185 CAS Number: 732251-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...