cis-2-Carbomethoxycyclopentane-1-carboxylic acid ≥95%
Supplier: Matrix Scientific
086429-5G
086429-1G
102848-634EA
1843.08
USD
102848-634
102848-638
cis-2-Carbomethoxycyclopentane-1-carboxylic acid ≥95%
cis-2-Carbomethoxycyclopentane-1-carboxylic acid
Hazard: Refrigerate
Formula:
C₈H₁₂O₄ Melting Pt: 41…42 °C |
MDL Number:
MFCD01311173 CAS Number: 92541-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...