cis-1,2-Cyclopentanedicarboxylic anhydride ≥97%

Supplier: Matrix Scientific

Synonyms: cis-Tetrahydro-1H-cyclopenta[c]furan-1,3(3aH)-dione

086425-5G
102848-612EA 512.31 USD
102848-612
cis-1,2-Cyclopentanedicarboxylic anhydride ≥97%
cis-1,2-Cyclopentanedicarboxylic anhydride

Hazard: Irritant

Formula: C₇H₈O₃
MW: 140.14 g/mol
Melting Pt: 67…71 °C
MDL Number: MFCD01311171
CAS Number: 35878-28-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR