Ethyl-3-bromothiophene-2-glyoxylate ≥97%
Supplier: Matrix Scientific
086418-5G
086418-1G
102848-568EA
530.77
USD
102848-568
102848-572
Ethyl-3-bromothiophene-2-glyoxylate ≥97%
Ethyl-3-bromothiophene-2-glyoxylate
Hazard: Irritant/Refrigerate
Formula:
C₈H₇BrO₃S |
MDL Number:
MFCD07782948 CAS Number: 70596-02-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...