Ethyl-2,4,5-trimethylbenzoylformate ≥97%

Supplier: Matrix Scientific

086417-2G
102848-564EA 827.69 USD
102848-564
Ethyl-2,4,5-trimethylbenzoylformate ≥97%
Ethyl-2,4,5-trimethylbenzoylformate

Hazard: Irritant

Formula: C₁₃H₁₆O₃
MDL Number: MFCD09801443
CAS Number: 80120-34-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR