Ethyl-6-methoxy-2-naphthoylformate ≥97%

Supplier: Matrix Scientific

086408-1G 086408-5G
102848-512EA 1792.31 USD
102848-512 102848-508
Ethyl-6-methoxy-2-naphthoylformate ≥97%
Ethyl-6-methoxy-2-naphthoylformate

Hazard: Irritant

Formula: C₁₅H₁₄O₄
MDL Number: MFCD09801433
CAS Number: 56547-13-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR