Ethyl-4-fluoro-1-naphthoylformate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(4-fluoronaphthalen-1-yl)-2-oxoacetate
086403-2G
102848-480EA
827.69
USD
102848-480
Ethyl-4-fluoro-1-naphthoylformate ≥97%
Ethyl-4-fluoro-1-naphthoylformate
Hazard: Irritant
Formula:
C₁₄H₁₁FO₃ MW: 246.24 g/mol |
MDL Number:
MFCD09801427 CAS Number: 409081-83-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...