Ethyl-2-fluoro-4-methylbenzoylformate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(2-fluoro-4-methylphenyl)-2-oxoacetate

086399-2G
102848-456EA 827.69 USD
102848-456
Ethyl-2-fluoro-4-methylbenzoylformate ≥97%
Ethyl-2-fluoro-4-methylbenzoylformate

Hazard: Irritant/Refrigerate

Formula: C₁₁H₁₁FO₃
MW: 210.21 g/mol
MDL Number: MFCD09801423
CAS Number: 873547-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR