Ethyl-3,5-dimethoxybenzoylformate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(3,5-dimethoxyphenyl)-2-oxoacetate
086382-2G
102848-354EA
1300
USD
102848-354
Ethyl-3,5-dimethoxybenzoylformate ≥97%
Ethyl-3,5-dimethoxybenzoylformate
Hazard: Irritant
Formula:
C₁₂H₁₄O₅ MW: 238.24 g/mol |
MDL Number:
MFCD09801406 CAS Number: 330551-16-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...