Ethyl-2,4-difluorobenzoylformate ≥97%

Supplier: Matrix Scientific

086378-2G
102848-330EA 827.69 USD
102848-330
Ethyl-2,4-difluorobenzoylformate ≥97%
Ethyl-2,4-difluorobenzoylformate

Hazard: Irritant

Formula: C₁₀H₈F₂O₃
MDL Number: MFCD06205165
CAS Number: 157372-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR