Ethyl-2-chlorobenzoylformate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(2-chlorophenyl)-2-oxoacetate

086363-1G 086363-5G
102848-238EA 606.15 USD
102848-238 102848-242
Ethyl-2-chlorobenzoylformate ≥97%
Ethyl-2-chlorobenzoylformate

Hazard: Irritant/Refrigerate

Formula: C₁₀H₉ClO₃
MW: 212.63 g/mol
MDL Number: MFCD09801392
CAS Number: 62123-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR