Ethyl-4-bromo-2-fluorobenzoylformate ≥97%

Supplier: Matrix Scientific

086359-1G 086359-5G
102848-214EA 530.77 USD
102848-214 102848-218
Ethyl-4-bromo-2-fluorobenzoylformate ≥97%
Ethyl-4-bromo-2-fluorobenzoylformate

Hazard: Irritant/Refrigerate

Formula: C₁₀H₈BrFO₃
MDL Number: MFCD09801388
CAS Number: 668970-56-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR