Ethyl-4-phenylbenzoylformate ≥97%

Supplier: Matrix Scientific

086358-2G
102848-210EA 616.92 USD
102848-210
Ethyl-4-phenylbenzoylformate ≥97%
Ethyl-4-phenylbenzoylformate

Hazard: Irritant

Formula: C₁₆H₁₄O₃
MDL Number: MFCD09801387
CAS Number: 6244-53-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR