Diethyl-2-oxononane-1,9-dicarboxylate ≥95%

Supplier: Matrix Scientific

086346-2G
102848-138EA 827.69 USD
102848-138
Diethyl-2-oxononane-1,9-dicarboxylate ≥95%
Diethyl-2-oxononane-1,9-dicarboxylate

Hazard: Irritant

Formula: C₁₃H₂₂O₅
MDL Number: MFCD01319632
CAS Number: 105600-21-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR