Diethyl-2-oxoheptane-1,7-dicarboxylate ≥95%

Supplier: Matrix Scientific

086344-2G
102848-126EA 827.69 USD
102848-126
Diethyl-2-oxoheptane-1,7-dicarboxylate ≥95%
Diethyl-2-oxoheptane-1,7-dicarboxylate

Hazard: Irritant/Refrigerate

Formula: C₁₁H₁₈O₅
MDL Number: MFCD01319630
CAS Number: 42212-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR