3-[(3,4-Ethylenedioxy)phenyl]-1-propene ≥97%

Supplier: Matrix Scientific

086288-2G
102847-802EA 769.23 USD
102847-802
3-[(3,4-Ethylenedioxy)phenyl]-1-propene ≥97%
3-[(3,4-Ethylenedioxy)phenyl]-1-propene

Hazard: Irritant

Formula: C₁₁H₁₂O₂
MDL Number: MFCD00626847
CAS Number: 58169-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR