3-(3,5-Dimethyl-4-methoxyphenyl)-1-propene ≥97%
Supplier: Matrix Scientific
086273-5G
086273-1G
102847-710EA
387.69
USD
102847-710
102847-714
3-(3,5-Dimethyl-4-methoxyphenyl)-1-propene ≥97%
3-(3,5-Dimethyl-4-methoxyphenyl)-1-propene
Hazard: Irritant
Formula:
C₁₂H₁₆O |
MDL Number:
MFCD09801270 CAS Number: 53483-16-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...