3-(3,4,5-Trimethoxyphenyl)-1-propene ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Allyl-1,2,3-trimethoxybenzene

086272-1G
102847-704EA 181.54 USD
102847-704
3-(3,4,5-Trimethoxyphenyl)-1-propene ≥97%
3-(3,4,5-Trimethoxyphenyl)-1-propene

Hazard: Irritant

Formula: C₁₂H₁₆O₃
MW: 208.26 g/mol
Boiling Pt: 106…108 °C (1 mmHg)
Density: 1.07
MDL Number: MFCD01656688
CAS Number: 487-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR