3-(3,5-Dichlorophenyl)-1-propene ≥97%

Supplier: Matrix Scientific

086269-2G
102847-688EA 560 USD
102847-688
3-(3,5-Dichlorophenyl)-1-propene ≥97%
3-(3,5-Dichlorophenyl)-1-propene

Hazard: Irritant

Formula: C₉H₈Cl₂
MDL Number: MFCD06201185
CAS Number: 75894-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR