4-(3,5-Dimethoxyphenyl)-1-butene ≥97%

Supplier: Matrix Scientific

086263-2G
102847-652EA 769.23 USD
102847-652
4-(3,5-Dimethoxyphenyl)-1-butene ≥97%
4-(3,5-Dimethoxyphenyl)-1-butene

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD09801196
CAS Number: 111540-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR