3-(3,5-Dimethoxyphenyl)-1-propene ≥97%

Supplier: Matrix Scientific

086262-5G 086262-1G
102847-644EA 387.69 USD
102847-644 102847-648
3-(3,5-Dimethoxyphenyl)-1-propene ≥97%
3-(3,5-Dimethoxyphenyl)-1-propene

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD09801195
CAS Number: 64118-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR