3-(2,6-Dimethoxyphenyl)-1-propene ≥97%

Supplier: Matrix Scientific

086257-2G
102847-616EA 560 USD
102847-616
3-(2,6-Dimethoxyphenyl)-1-propene ≥97%
3-(2,6-Dimethoxyphenyl)-1-propene

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MDL Number: MFCD09801187
CAS Number: 3698-35-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR