3-(2,5-Dimethoxyphenyl)-2-methyl-1-propene ≥97%

Supplier: Matrix Scientific

086255-2G
102847-604EA 560 USD
102847-604
3-(2,5-Dimethoxyphenyl)-2-methyl-1-propene ≥97%
3-(2,5-Dimethoxyphenyl)-2-methyl-1-propene

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MDL Number: MFCD09801185
CAS Number: 35205-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR