4-(4-Ethoxyphenyl)-2-methyl-1-butene ≥97%

Supplier: Matrix Scientific

086242-2G
102847-532EA 769.23 USD
102847-532
4-(4-Ethoxyphenyl)-2-methyl-1-butene ≥97%
4-(4-Ethoxyphenyl)-2-methyl-1-butene

Hazard: Irritant

Formula: C₁₃H₁₈O
MDL Number: MFCD09801138
CAS Number: 18272-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR