3-(4-Acetoxyphenyl)-2-methyl-1-propene ≥97%

Supplier: Matrix Scientific

086230-2G
102847-460EA 560 USD
102847-460
3-(4-Acetoxyphenyl)-2-methyl-1-propene ≥97%
3-(4-Acetoxyphenyl)-2-methyl-1-propene

Hazard: Irritant

Formula: C₁₂H₁₄O₂
MDL Number: MFCD07698674
CAS Number: 890097-89-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR