4-(2-Acetoxyphenyl)-1-butene ≥97%

Supplier: Matrix Scientific

086224-2G
102847-424EA 769.23 USD
102847-424
4-(2-Acetoxyphenyl)-1-butene ≥97%
4-(2-Acetoxyphenyl)-1-butene

Hazard: Irritant

Formula: C₁₂H₁₄O₂
MDL Number: MFCD07782970
CAS Number: 890097-68-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR