4-(3-Methoxyphenyl)-2-methyl-1-butene ≥97%

Supplier: Matrix Scientific

086220-2G
102847-400EA 769.23 USD
102847-400
4-(3-Methoxyphenyl)-2-methyl-1-butene ≥97%
4-(3-Methoxyphenyl)-2-methyl-1-butene

Hazard: Irritant

Formula: C₁₂H₁₆O
MDL Number: MFCD07698649
CAS Number: 40463-03-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR