3-(2-Methoxyphenyl)-2-methyl-1-propene ≥97%
Supplier: Matrix Scientific
086215-5G
086215-1G
102847-368EA
387.69
USD
102847-368
102847-372
3-(2-Methoxyphenyl)-2-methyl-1-propene ≥97%
3-(2-Methoxyphenyl)-2-methyl-1-propene
Hazard: Irritant
Formula:
C₁₁H₁₄O |
MDL Number:
MFCD07698642 CAS Number: 91969-32-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...