3-(4-Chlorophenyl)-1-propene ≥97%
Supplier: Matrix Scientific
Synonyms:
1-Allyl-4-chlorobenzene
086188-1G
086188-5G
086188-2G
102847-206EA
620
USD
102847-206
102847-208
102847-210
3-(4-Chlorophenyl)-1-propene ≥97%
3-(4-Chlorophenyl)-1-propene
Hazard: Irritant
Formula:
C₉H₉Cl MW: 152.63 g/mol Boiling Pt: 199…201 °C |
MDL Number:
MFCD01319547 CAS Number: 1745-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...