Ethyl-5-methyl-5-hexenoate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-methylhex-5-enoate

086175-5G 086175-1G 086175-2G
102847-128EA 415.38 USD
102847-128 102847-132 102847-130
Ethyl-5-methyl-5-hexenoate ≥97%
Ethyl-5-methyl-5-hexenoate

Hazard: Irritant

Formula: C₉H₁₆O₂
MW: 156.23 g/mol
MDL Number: MFCD00671855
CAS Number: 39495-82-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR