2-Chloro-N,N-dimethylacetamide ≥97%
Supplier: Matrix Scientific
086134-1G
102847-060EA
383.08
USD
102847-060
2-Chloro-N,N-dimethylacetamide ≥97%
2-Chloro-N,N-dimethylacetamide
Hazard: Irritant
Formula:
C₄H₈ClNO |
MDL Number:
MFCD00039360 CAS Number: 2675-89-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...