Methyl-4-chlorobutanimidoate hydrochloride ≥97%

Supplier: Matrix Scientific

086130-1G
102847-048EA 378.46 USD
102847-048
Methyl-4-chlorobutanimidoate hydrochloride ≥97%
Methyl-4-chlorobutanimidoate hydrochloride

Hazard: Irritant

Formula: C₅H₁₁Cl₂NO
MDL Number: MFCD12404920
CAS Number: 77570-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR