N-Ethyl-N-methylguanidine ≥97%

Supplier: Matrix Scientific

086122-1G
102847-016EA 553.85 USD
102847-016
N-Ethyl-N-methylguanidine ≥97%
N-Ethyl-N-methylguanidine

Hazard: Irritant

Formula: C₄H₁₁N₃
MDL Number: MFCD08741472
CAS Number: 1113-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR