Triisopropylchlorosilane ≥97%
Supplier: Matrix Scientific
Synonyms:
Chlorotriisopropylsilane
, Triisopropylsilyl chloride
, Triisopropylchlorosilane
086113-100G
086113-25G
102846-978EA
60
USD
102846-978
102846-980
Triisopropylchlorosilane ≥97%
Triisopropylchlorosilane
Hazard: Corrosive/Moisture Sensitive
Formula:
C₉H₂₁ClSi MW: 192.8 g/mol Boiling Pt: 200 °C Density: 0.908 Flash Pt: 62 °C |
MDL Number:
MFCD00009656 CAS Number: 13154-24-0 UN: 2987 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...