2-Cyano-N-ethylacetamide ≥97%

Supplier: Matrix Scientific

086112-1G
102846-976EA 524.62 USD
102846-976
2-Cyano-N-ethylacetamide ≥97%
2-Cyano-N-ethylacetamide

Hazard: Irritant

Formula: C₅H₈N₂O
MDL Number: MFCD00118415
CAS Number: 15029-36-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR