Pivaloylacetonitrile ≥97%

Supplier: Matrix Scientific

Synonyms: 4,4-Dimethyl-3-oxovaleronitrile4,4-Dimethyl-3-oxopentanenitrileTrimethylacetylacetonitrile

086106-50G 086106-10G
102846-956EA 53.85 USD
102846-956 102846-958
Pivaloylacetonitrile ≥97%
Pivaloylacetonitrile

Purity: 97%
CAS: 59997-51-2
MDL Number: MFCD00010208
Molecular Formula: C7H11NO
Molecular Weight: 125,17
Hazard: Irritant

Formula: C₇H₁₁NO
MW: 125.17 g/mol
MDL Number: MFCD00010208
CAS Number: 59997-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR