N-Methylhydrazinecarboximidamide hydroiodide ≥97%

Supplier: Matrix Scientific

086098-1G 086098-10G
102846-926EA 340 USD
102846-926 102846-928
N-Methylhydrazinecarboximidamide hydroiodide ≥97%
N-Methylhydrazinecarboximidamide hydroiodide

Hazard: Irritant

Formula: C₂H₉IN₄
MW: 216.03 g/mol
MDL Number: MFCD00179313
CAS Number: 33398-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR