N-Methyl-N-prop-2-ynylamine ≥95%

Supplier: Matrix Scientific

Synonyms: N-Methylpropargylamine

086096-10G 086096-50G 086096-1G
102846-920EA 3369.23 USD
102846-920 102846-916 102846-918
N-Methyl-N-prop-2-ynylamine ≥95%
N-Methyl-N-prop-2-ynylamine

Hazard: Irritant

Formula: C₄H₇N
MW: 69.11 g/mol
MDL Number: MFCD00008573
CAS Number: 35161-71-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR