Triethylphosphine sulfide ≥97%
Supplier: Matrix Scientific
086072-1G
102846-842EA
323.08
USD
102846-842
Triethylphosphine sulfide ≥97%
Triethylphosphine sulphide
Hazard: Irritant
Formula:
C₆H₁₅PS |
MDL Number:
MFCD00014462 CAS Number: 597-51-3 UN: 3464 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...