1-Nitropropan-2-ol ≥97%

Supplier: Matrix Scientific

086070-1G
102846-838EA 806.15 USD
102846-838
1-Nitropropan-2-ol ≥97%
1-Nitropropan-2-ol

Hazard: Irritant

Formula: C₃H₇NO₃
MDL Number: MFCD00506013
CAS Number: 3156-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR