3-Ethylacetylacetone ≥97%
Supplier: Matrix Scientific
Synonyms:
3-Ethyl-2,4-pentanedione
, 3-Ethyl-4-hydroxy-3-penten-3-one
086065-1G
102846-820EA
212.31
USD
102846-820
3-Ethylacetylacetone ≥97%
3-Ethylacetylacetone
Hazard: Irritant
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol |
MDL Number:
MFCD00008764 CAS Number: 1540-34-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...