L-Threoninamide hydrochloride ≥97%
Supplier: Matrix Scientific
Synonyms:
H-Thr-NH₂·HCl
, (2S,3R)-2-Amino-3-hydroxybutanamide hydrochloride
086061-1G
086061-10G
102846-804EA
30.77
USD
102846-804
102846-806
L-Threoninamide hydrochloride ≥97%
L-Threoninamide hydrochloride
Hazard: Irritant
Formula:
C₄H₁₁ClN₂O₂ MW: 154.594 g/mol |
MDL Number:
MFCD00058288 CAS Number: 33209-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...