N-Benzyl-N,N-dimethyl-3-(lauroylamino)propan-1-aminium chloride ≥97%

Supplier: Matrix Scientific

086047-1G
102846-756EA 920 USD
102846-756
N-Benzyl-N,N-dimethyl-3-(lauroylamino)propan-1-aminium chloride ≥97%
N-Benzyl-N,N-dimethyl-3-(lauroylamino)propan-1-aminium chloride

Hazard: Irritant

Formula: C₂₄H₄₃ClN₂O
MW: 411.08 g/mol
MDL Number: MFCD18089285
CAS Number: 52513-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR