Diphenyl-2-pyridylphosphine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-(Diphenylphosphino)pyridine

086032-10G 086032-1G
102846-700EA 4998.56 USD
102846-700 102846-702
Diphenyl-2-pyridylphosphine ≥97%
Diphenyl-2-pyridylphosphine

Purity: 97%
CAS: 37943-90-1
MDL Number: MFCD00192108
Molecular Formula: C17H14NP
Molecular Weight: 263,28
Hazard: Irritant

Formula: C₁₇H₁₄NP
MW: 263.28 g/mol
Melting Pt: 82…84 °C
MDL Number: MFCD00192108
CAS Number: 37943-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR